Invitrogen
Manufacturer Code:PA5117302
Catalog # PIPA5117302
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant protein encompassing a sequence within the center region of human COL4A3BP. The exact sequence is proprietary. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 2810404O15Rik 9230101K08Rik alpha 3 type IV collagen binding protein AU016711 ceramide transfer protein ceramide transport protein ceramide transporter CERT CERTL Col4a3bp collagen type IV alpha 3 binding protein collagen type IV alpha-3-binding protein collagen type IV alpha 3 (Goodpasture antigen) binding protein goodpasture antigen-binding protein GPBP hCERT lipid-transfer protein CERTL MRD34 procollagen type IV alpha 3 (Goodpasture antigen) binding protein STARD11 StAR-related lipid transfer (START) domain containing 11 stAR-related lipid transfer protein 11 START domain-containing protein 11; Ceramide transfer protein; ceramide transporter; Collagen type IV alpha-3-binding protein; collagen, type IV, alpha 3 (Goodpasture antigen) binding protein; Goodpasture antigen-binding protein; GPBP; hCERT; lipid-transfer protein CERTL; StAR-related lipid transfer (START) domain containing 11; StAR-related lipid transfer protein 11; StARD11; START domain-containing protein 11
Gene Aliases: CERT; CERT1; CERTL; COL4A3BP; GPBP; MRD34; STARD11
UniProt ID: (Human) Q9Y5P4
Entrez Gene ID: (Human) 10087
Molecular Function:
kinase
protein kinase
transfer/carrier protein
transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.