Invitrogen
Manufacturer Code:PA129485
Catalog # PIPA129485
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Chicken |
Class | Polyclonal |
Type | Antibody |
Immunogen | synthetic peptide VVEEGEMKVY RREVEENGIV LDPLKATHAV KGVTGHEVCN YFWNVDVRND WETTIENFHV VETLADNAII IYQTHKRVWP ASQRDVLYLS VIRKIPALTE NDPETWIVCN FSVDHDSAPL NNRCVRAKIN VAMICQTLVS PPEGNQEISR DNILCKITYV ANVNPGGWAP ASVLRAVA corresponding to C terminal amino acid |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C,Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles.,avoiding freeze/thaw cycles. |
Chicken Polyclonal Antibody
For Research Use Only
Protein Aliases: alpha 3 type IV collagen binding protein; Ceramide transfer protein; ceramide transport protein; ceramide transporter; CERT; CERT CERTL FLJ20597 GPBP STARD11; Collagen type IV alpha-3-binding protein; collagen, type IV, alpha 3 (Goodpasture antigen) binding protein; Goodpasture antigen-binding protein; GPBP; hCERT; lipid-transfer protein CERTL; procollagen, type IV, alpha 3 (Goodpasture antigen) binding protein; StAR-related lipid transfer (START) domain containing 11; StAR-related lipid transfer protein 11; StARD11; START domain-containing protein 11
Gene Aliases: 2810404O15Rik; 9230101K08Rik; AU016711; CERT; CERT1; CERTL; COL4A3BP; GPBP; MRD34; STARD11
UniProt ID: (Human) Q9Y5P4, (Mouse) Q9EQG9
Entrez Gene ID: (Human) 10087, (Mouse) 68018, (Rat) 365652
Molecular Function: kinase protein kinase transfer/carrier protein transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.