Novus Biologicals
Manufacturer Code:NBP106028
Catalog # NBP106028
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Goat |
Class | Polyclonal |
Type | Antibody |
Immunogen | Peptide with sequence C-KREDSWQKRLDKETEK corresponding to internal region according to NP_005704.1 NP_112729.1. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20C. Avoid freeze-thaw cycles. |
Goat Polyclonal Antibody
For Research Use Only
Protein Aliases: Ceramide transfer protein; Ceramide transfer protein CERTFLJ20597 CERTL collagen type IV alpha-3-binding protein collagen type IV alpha 3 (Goodpasture antigen) binding protein Goodpasture antigen-binding protein GPBPSTARD11ceramide transporter hCERT lipid-transfer protein CERTL StARD11 StAR-related lipid transfer (START) domain containing 11 StAR-related lipid transfer protein 11 START domain containing 11 START domain-containing protein 11; ceramide transporter; Collagen type IV alpha-3-binding protein; collagen, type IV, alpha 3 (Goodpasture antigen) binding protein; Goodpasture antigen-binding protein; GPBP; hCERT; lipid-transfer protein CERTL; StAR-related lipid transfer (START) domain containing 11; StAR-related lipid transfer protein 11; StARD11; START domain-containing protein 11
Gene Aliases: CERT; CERT1; CERTL; COL4A3BP; GPBP; MRD34; STARD11
UniProt ID: (Human) Q9Y5P4
Entrez Gene ID: (Human) 10087
Molecular Function: kinase protein kinase transfer/carrier protein transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.