Invitrogen
Manufacturer Code:PA585634
Catalog # PIPA585634
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit protein encompassing a sequence within the N-terminus of human COL4A1. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: alpha1(IV) collagen; Arresten; COL4A1 NC1 domain; Collagen alpha-1(IV) chain; collagen IV, alpha-1 polypeptide; collagen of basement membrane, alpha-1 chain; collagen, type IV, alpha 1; Del(8)Bru44H; POREN1 Antibody collagen type IV alpha 1 Antibody ICH Antibody HANAC Antibody arresten Antibody; procollagen, type IV, alpha 1; retinal anterior wiring
Gene Aliases: Bru; BSVD; Col4a-1; COL4A1; Del(8)44H; RATOR; Raw; Svc
UniProt ID: (Human) P02462, (Mouse) P02463
Entrez Gene ID: (Human) 1282, (Mouse) 12826
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.