Abnova
Manufacturer Code:PAB3065
Catalog # 89-130-808
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide (conjugated with KLH) corresponding to C-terminus of human COL2A1. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C. For long term storage store at -20°C.<br />Aliquot to avoid repeated freezing and thawing. |
Rabbit polyclonal antibody raised against synthetic peptide of COL2A1.
For Research Use Only
Protein Aliases: Alpha-1 type II collagen; ANFH; AOM; arthroophthalmopathy, progressive (Stickler syndrome); cartilage collagen; Chondrocalcin; COL11A3; Collagen alpha-1(II) chain; collagen II, alpha-1 polypeptide; collagen, type II, alpha 1; MGC131516; SEDC
Gene Aliases: ANFH; AOM; COL11A3; COL2A1; SEDC; STL1
UniProt ID: (Human) P02458
Entrez Gene ID: (Human) 1280
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.