Abnova
Manufacturer Code:PAB13494
Catalog # 89-116-703
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (PFA fixed) (IHC (PFX)) | Assay dependent |
Radioimmune Assays (RIA) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Native purified COL2A1 from bovine cartilage. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C.<br />After reconstitution with 100 uL of deionized water store at -20°C.<br />Aliquot to avoid repeated freezing and thawing. |
Rabbit polyclonal antibody raised against native COL2A1.
For Research Use Only
Protein Aliases: Alpha-1 type II collagen; CB11; Collagen alpha-1(II) chain; collagen, type II, alpha 1; pro-alpha 1(II); unnamed protein product
Gene Aliases: CB11; COL2A1
UniProt ID: (Bovine) P02459
Entrez Gene ID: (Bovine) 407142
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.