Invitrogen
Manufacturer Code:PA529569
Catalog # PIPA529569
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit fragment corresponding to a region within amino acids 219 and 478 of Human COL1A1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: alpha-1 type 1 collagen; Alpha-1 type I collagen; alpha1(I) procollagen; collagen alpha 1 chain type I; Collagen alpha-1(I) chain; collagen alpha-1(I) chain preproprotein; collagen of skin, tendon and bone, alpha-1 chain; collagen, type I, alpha 1; OI4 alpha-1 type I collagen collagen alpha 1 chain type I collagen alpha-1(I) chain collagen of skin tendon and bone alpha-1 chain pro-alpha-1 collagen type 1; pro-alpha-1 collagen type 1; procollagen, type I, alpha 1; type I proalpha 1; type I procollagen alpha 1 chain
Gene Aliases: Col1a-1; COL1A1; Cola-1; Cola1; EDSC; Mov-13; Mov13; OI1; OI2; OI3; OI4
UniProt ID: (Human) P02452, (Mouse) P11087
Entrez Gene ID: (Human) 1277, (Mouse) 12842
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.