Abnova
Manufacturer Code:PAB17519
Catalog # 89-119-180
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide corresponding to internal of human COL13A1. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C.<br />Aliquot to avoid repeated freezing and thawing. |
Rabbit polyclonal antibody raised against synthetic peptide of COL13A1.
For Research Use Only
Protein Aliases: Collagen alpha-1(XIII) chain; collagen, type XIII, alpha 1; COLXIIIA1; FLJ42485
Gene Aliases: CMS19; COL13A1; COLXIIIA1
UniProt ID: (Human) Q5TAT6
Entrez Gene ID: (Human) 1305
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.