Abnova
Manufacturer Code:PAB31414
Catalog # 89-998-259
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant protein corresponding to human COL12A1. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: BA209D8.1; COL12A1L; Collagen alpha-1(XII) chain; collagen type XII proteoglycan; collagen, type XII, alpha 1; DJ234P15.1
Gene Aliases: BA209D8.1; BTHLM2; COL12A1; COL12A1L; DJ234P15.1; UCMD2
UniProt ID: (Human) Q99715
Entrez Gene ID: (Human) 1303
Molecular Function: receptor transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.