Invitrogen
Manufacturer Code:PA568410
Catalog # PIPA568410
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Fusion protein of human COL11A1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: COBA1 COL11A1 COLL6 Collagen alpha 1 Collagen alpha-1(XI) chain collagen XI alpha 1 collagen XI alpha 1 polypeptide collagen type XI alpha 1 STL2 STL3 XI chain precursor; Collagen alpha-1(XI) chain; collagen XI, alpha-1 polypeptide; collagen, type XI, alpha 1
Gene Aliases: CO11A1; COL11A1; COLL6; STL2
UniProt ID: (Human) P12107
Entrez Gene ID: (Human) 1301
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.