Novus Biologicals
Manufacturer Code:NBP19846320UL
Catalog # NBP19846320
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Mouse |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | The immunogen for this antibody is Col11a1 - middle region. Peptide sequence AGPPGPPGKRGPPGASGSEGRQGEKGAKGEAGAEGPPGKTGPVGPQGPSG. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: alpha-1 polypeptide COLL6 collagen type XI alpha 1; Collagen alpha-1(XI) chain; collagen XI, alpha-1 polypeptide; collagen, type XI, alpha 1
Gene Aliases: CO11A1; COL11A1; COLL6; STL2
UniProt ID: (Human) P12107
Entrez Gene ID: (Human) 1301
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.