Invitrogen
Manufacturer Code:PA529044
Catalog # PIPA529044
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit fragment corresponding to a region within amino acids 7 and 286 of Human COG7 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: CDG2E COG complex subunit 7 conserved oligomeric Golgi complex subunit 7; COG complex subunit 7; Component of oligomeric Golgi complex 7; Conserved oligomeric Golgi complex subunit 7
Gene Aliases: CDG2E; COG7; UNQ3082/PRO10013
UniProt ID: (Human) P83436
Entrez Gene ID: (Human) 91949
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.