Proteintech
Manufacturer Code:135521AP150UL
Catalog # 50-172-8829
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | COG7 Fusion Protein Ag4335 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: CDG2E COG complex subunit 7 COG7; COG complex subunit 7; Component of oligomeric Golgi complex 7; Conserved oligomeric Golgi complex subunit 7
Gene Aliases: 5630400E24Rik; CDG2E; COG7; Gm167; UNQ3082/PRO10013
UniProt ID: (Human) P83436, (Mouse) Q3U3L7, (Rat) Q3T1G7
Entrez Gene ID: (Human) 91949, (Mouse) 233824, (Rat) 293456
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.