Invitrogen
Manufacturer Code:PA554169
Catalog # PIPA554169
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit protein corresponding to Human COBRA1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Cofactor of BRCA1; Negative elongation factor B; negative elongation factor complex member B, Cobra1; negative elongation factor protein B; NELF-B; ortholog of human cofactor of BRCA1 (COBRA1); RP13-122B23.3 DKFZp586B0519 KIAA1182 NELF-B NELFB negative elongation factor B negative elongation factor protein B
Gene Aliases: A730008L03Rik; AB041607; AI663983; COBRA1; KIAA1182; MNCb-5210; NELF-B; NELFB; RGD1307832
UniProt ID: (Human) Q8WX92, (Mouse) Q8C4Y3
Entrez Gene ID: (Human) 25920, (Mouse) 58202, (Rat) 311796
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.