Invitrogen
Manufacturer Code:PA5115638
Catalog # PIPA5115638
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthesized peptide derived from human NELFB corresponding to a region within C-terminal amino acids. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: A730008L03Rik AB041607 AI663983 COBRA1 Cofactor of BRCA1 DKFZp586B0519 KIAA1182 MNCb-5210 Negative elongation factor B negative elongation factor complex member B negative elongation factor complex member B Cobra1 negative elongation factor protein B NELFB NELF-B ortholog of human cofactor of BRCA1 (COBRA1) RGD1307832 RP13-122B23.3; Cofactor of BRCA1; Negative elongation factor B; negative elongation factor complex member B, Cobra1; negative elongation factor protein B; NELF-B; ortholog of human cofactor of BRCA1 (COBRA1)
Gene Aliases: A730008L03Rik; AB041607; AI663983; COBRA1; KIAA1182; MNCb-5210; NELF-B; NELFB; RGD1307832
UniProt ID: (Human) Q8WX92, (Mouse) Q8C4Y3
Entrez Gene ID: (Human) 25920, (Mouse) 58202, (Rat) 311796
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.