Novus Biologicals
Manufacturer Code:NBP105977
Catalog # NBP105977
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | The immunogen recognized by this antibody maps to a region between residue 530 and 580 of human cofactor of BRCA1 using the numbering given in entry NP_056271.2 (GeneID 25920). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C. Do not freeze. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Cofactor of BRCA1; cofactor of BRCA1DKFZp586B0519 KIAA1182negative elongation factor B NELFB NELF-Bnegative elongation factor protein B; Negative elongation factor B; negative elongation factor protein B; NELF-B
Gene Aliases: COBRA1; KIAA1182; NELF-B; NELFB
UniProt ID: (Human) Q8WX92
Entrez Gene ID: (Human) 25920
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.