Novus Biologicals
Manufacturer Code:NBP170505
Catalog # NBP170505
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptides corresponding to COBLL1(COBL-like 1) The peptide sequence was selected from the middle region of COBLL1. Peptide sequence QAKPSSFFLQMQKRVSGHYVTSAAAKSVHAAPNPAPKELTNKEAERDMLP. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: COBL-like 1; COBL-like 1 COBLR1 cordon-bleu protein-like 1; Cordon-bleu protein-like 1; cordon-bleu WH2 repeat protein-like 1
Gene Aliases: COBLL1; COBLR1; KIAA0977
UniProt ID: (Human) Q53SF7
Entrez Gene ID: (Human) 22837
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.