Abnova
Manufacturer Code:MAB11916
Catalog # 89-110-053
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | S67-25 |
Immunogen | Recombinant fusion protein corresponding to amino acids of 96-1265 at extracellular domain of human CNTNAP2. |
Conjugate | ATTO 700 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C.<br />Aliquot to avoid repeated freezing and thawing. |
Mouse monoclonal antibody raised against partial recombinant CNTNAP2.
For Research Use Only
Protein Aliases: AUTS15; CASPR2; CDFE; Cell recognition molecule Caspr2; Contactin-associated protein-like 2; DKFZp781D1846; homolog of Drosophila neurexin IV; NRXN4
Gene Aliases: AUTS15; CASPR2; CDFE; CNTNAP2; KIAA0868; NRXN4; PTHSL1
UniProt ID: (Human) Q9UHC6
Entrez Gene ID: (Human) 26047
Molecular Function: apolipoprotein cell adhesion molecule enzyme modulator extracellular matrix protein hydrolase membrane-bound signaling molecule metalloprotease oxidase oxidoreductase protease receptor serine protease signaling molecule transfer/carrier protein transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.