Invitrogen
Manufacturer Code:PA5115032
Catalog # PIPA5115032
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthesized peptide derived from human CNTNAP1 corresponding to a region within N-terminal amino acids. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: AI841080 Caspr caspr1 CNTNAP CNTNAP1 contactin associated protein 1 contactin associated protein-like 1 contactin-associated protein 1 MHDNIV NCP1 neurexin 4 neurexin 4 (contactin associated protein) neurexin IV Neurexin-4 Nrxn4 p190 Paranodin shm; Caspr; caspr1; contactin associated protein 1; Contactin-associated protein 1; MHDNIV; NCP1; neurexin 4 (contactin associated protein); Neurexin IV; Neurexin-4; p190; Paranodin
Gene Aliases: AI841080; CASPR; CNTNAP; CNTNAP1; NCP1; NRXN4; P190; shm
UniProt ID: (Human) P78357, (Mouse) O54991
Entrez Gene ID: (Human) 8506, (Mouse) 53321
Molecular Function: apolipoprotein cell adhesion molecule enzyme modulator extracellular matrix protein hydrolase membrane-bound signaling molecule metalloprotease oxidase oxidoreductase protease receptor serine protease signaling molecule transfer/carrier protein transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.