Abnova
Manufacturer Code:H00008506A01
Catalog # 89-105-873
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Polyclonal |
Type | Antibody |
Immunogen | 22 a.a. to 131 a.a) partial recombinant protein with GST tag.,CNTNAP1 (NP_003623 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Mouse polyclonal antibody raised against a partial recombinant CNTNAP1.
For Research Use Only
Protein Aliases: CASPR; caspr1; CNTNAP; Contactin-associated protein 1; Neurexin IV; Neurexin-4; NRXN4; P190
Gene Aliases: CASPR; CNTNAP; CNTNAP1; NRXN4; P190
UniProt ID: (Human) P78357
Entrez Gene ID: (Human) 8506
Molecular Function: apolipoprotein cell adhesion molecule enzyme modulator extracellular matrix protein hydrolase membrane-bound signaling molecule metalloprotease oxidase oxidoreductase protease receptor serine protease signaling molecule transfer/carrier protein transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.