Invitrogen
Manufacturer Code:PA547437
Catalog # PIPA547437
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Neutralization (Neu) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Goat |
Class | Polyclonal |
Type | Antibody |
Immunogen | Sf 21-derived recombit human CNTF R alpha Gln23-Pro346 (predicted) |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Goat Polyclonal Antibody
For Research Use Only
Protein Aliases: Ciliary neurotrophic factor receptor subunit alpha; CNTF receptor subunit alpha; CNTFR ciliary neurotrophic factor receptor MGC1774 CNTFR alpha ciliary neurotrophic factor receptor alpha precursor; CNTFR-alpha
Gene Aliases: CNTFR
UniProt ID: (Human) P26992
Entrez Gene ID: (Human) 1271
Molecular Function: cytokine defense/immunity protein signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.