Invitrogen
Manufacturer Code:PA545053
Catalog # PIPA545053
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide directed towards the C-terminal of human CNTFR |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: ciliary neurotrophic factor receptor alpha; Ciliary neurotrophic factor receptor MGC1774; Ciliary neurotrophic factor receptor subunit alpha; CNTF receptor subunit alpha; CNTFR alpha; CNTFR-alpha
Gene Aliases: CNTFR; CNTFRALPHA
UniProt ID: (Dog) Q71DR4, (Human) P26992, (Rat) Q08406, (Mouse) O88507
Entrez Gene ID: (Sheep) 100885760, (Dog) 442941, (Bovine) 539548, (Horse) 100146590, (Rabbit) 100349367, (Human) 1271, (Rat) 313173, (Mouse) 12804, (Guinea pig) 100722634
Molecular Function:
cytokine
defense/immunity protein
signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.