Novus Biologicals
Manufacturer Code:NBP16894320UL
Catalog # NBP16894320
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptides corresponding to CNTFR (ciliary neurotrophic factor receptor) The peptide sequence was selected from the C terminal of CNTFR. Peptide sequence VAAHATPWTEEPRHLTTEAQAAETTTSTTSSLAPPPTTKICDPGELGSGG. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: ciliary neurotrophic factor receptor ciliary neurotrophic factor receptor alpha ciliary neurotrophic factor receptor subunit alpha CNTF receptor subunit alpha CNTFR alpha CNTFR-alpha MGC1774; Ciliary neurotrophic factor receptor subunit alpha; CNTF receptor subunit alpha; CNTFR-alpha
Gene Aliases: CNTFR
UniProt ID: (Human) P26992
Entrez Gene ID: (Human) 1271
Molecular Function: cytokine defense/immunity protein signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.