Invitrogen
Manufacturer Code:PA579055
Catalog # PIPA579055
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | E.coli-derived mouse CNTF recombit protein (Position: A2-M198). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Ciliary neurotrophic factor; Ciliary neurotrophic factor CNTF Cntf; ciliary neurotropic factor; CNTF
Gene Aliases: AI429687; Cntf
UniProt ID: (Mouse) P51642, (Rat) P20294
Entrez Gene ID: (Mouse) 12803, (Rat) 25707
Molecular Function:
growth factor
neurotrophic factor
signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.