Abnova
Manufacturer Code:PAB26137
Catalog # 89-127-778
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (PFA fixed) (IHC (PFX)) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide corresponding to 19 amino acids at internal region of human CNR2. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C. For long term storage store at -80°C.<br />Aliquot to avoid repeated freezing and thawing. |
Rabbit polyclonal antibody raised against synthetic peptide of CNR2.
For Research Use Only
Protein Aliases: Cannabinoid receptor 2; cannabinoid receptor 2 (macrophage); CB-2; CB2; CX5; testis-dominant CNR2 isoform CB2
Gene Aliases: CB-2; CB2; CB2A; CB2B; CNR2; CX5
UniProt ID: (Human) P34972
Entrez Gene ID: (Human) 1269
Molecular Function: G-protein coupled receptor receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.