Proteintech
Manufacturer Code:667291IG150UL
Catalog # 50-173-7141
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 1B9G9 |
Immunogen | CNPase Fusion Protein Ag4227 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: 2', 3' cyclic nucleotide 3' phosphohydrolase; 2',3'-cyclic-nucleotide 3'-phosphodiesterase; CNP; CNP CNP1 CNPase; CNPase
Gene Aliases: CNP; CNP1; CNPF; CNPI; CNPII
UniProt ID: (Human) P09543, (Rat) P13233
Entrez Gene ID: (Human) 1267, (Pig) 100511587, (Rat) 25275
Molecular Function:
hydrolase
phosphodiesterase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.