Invitrogen
Manufacturer Code:PA595392
Catalog # PIPA595392
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide corresponding to a sequence in the middle region of human CNPase identical to the related mouse sequence and different from the related rat sequence by one amino acid. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 2&prime3&prime-cyclic-nucleotide 3&prime-phosphodiesterase CNP CNPase 3.1.4.37 CNP; 2', 3' cyclic nucleotide 3' phosphohydrolase; 2',3'-cyclic-nucleotide 3'-phosphodiesterase; CNP; CNPase; cyclic nucleotide phosphodiesterase 1
Gene Aliases: CNP; Cnp-1; CNP1; CNPase; CNPF; CNPI; CNPII
UniProt ID: (Human) P09543, (Mouse) P16330, (Rat) P13233
Entrez Gene ID: (Human) 1267, (Mouse) 12799, (Rat) 25275
Molecular Function:
hydrolase
phosphodiesterase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.