Novus Biologicals
Manufacturer Code:NBP185995
Catalog # NBP185995
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | This antibody was developed against Recombinant Protein corresponding to amino acids:TLARVIVDKYRDGTKMVSADAYKITPGARGAFSEEYKRLDEDLAAYCRRRDIRILVLDDTNHERERLEQLFEMADQYQYQVVLVEPKTAWRLDCAQLKEKNQW |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 2' 3' cyclic nucleotide 3' phosphohydrolase 2'-3'-cyclic nucleotide 3' phosphodiesterase 2'-3'-cyclic-nucleotide 3'-phosphodiesterase CNP1 CNPase EC 3.1.4.37; 2', 3' cyclic nucleotide 3' phosphohydrolase; 2',3'-cyclic-nucleotide 3'-phosphodiesterase; CNP; CNPase
Gene Aliases: CNP; CNP1
UniProt ID: (Human) P09543
Entrez Gene ID: (Human) 1267
Molecular Function:
hydrolase
phosphodiesterase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.