R&D Systems
Manufacturer Code:MAB3739
Catalog # MAB3739
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rat |
Class | Monoclonal |
Type | Antibody |
Clone | 347625 |
Immunogen | <EM>E. coli</EM>-derived recombinant human CNP Lys24-Cys126 Accession # P23582 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt -20 to -70 degreesC as supplied. 1 month 2 to 8 degreesC under sterile conditions after reconstitution. 6 months -20 to -70 degreesC under sterile con |
Rat Monoclonal Antibody
For Research Use Only
Protein Aliases: C-type natriuretic peptide; CNP CNP2 C-type natriuretic peptide natriuretic peptide C natriuretic peptide precursor C NPPC; CNP-22; CNP-29; CNP-53; natriuretic peptide precursor type C
Gene Aliases: CNP; CNP2; NPPC
UniProt ID: (Human) P23582
Entrez Gene ID: (Human) 4880
Molecular Function: peptide hormone signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.