Invitrogen
Manufacturer Code:MA524085
Catalog # PIMA524085
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rat |
Class | Monoclonal |
Type | Antibody |
Clone | 347625 |
Immunogen | E. coli-derived recombit human CNP Lys24-Cys126 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rat Monoclonal Antibody
For Research Use Only
Protein Aliases: C-type natriuretic peptide; CNP-22; CNP-29; CNP-53; natriuretic peptide precursor type C; NPPC CNP CNP2 C-type natriuretic peptide natriuretic peptide C natriuretic peptide precursor C
Gene Aliases: CNP; CNP2; NPPC
UniProt ID: (Human) P23582
Entrez Gene ID: (Human) 4880
Molecular Function: peptide hormone signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.