Invitrogen
Manufacturer Code:PA5103119
Catalog # PIPA5103119
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthesized peptide derived from human CNP-53 corresponding to a region within the internal amino acids. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: C-type natriuretic peptide; Cnp CNP2 CNP-22 CNP-29 CNP-53 C-type natriuretic peptide INT3 lbab natriuretic peptide C natriuretic peptide precursor C natriuretic peptide precursor type C natriuretic peptide type C NOTCH3 NPPC; CNP-22; CNP-29; CNP-53; natriuretic peptide precursor C; natriuretic peptide precursor type C
Gene Aliases: CNP; CNP2; lbab; NPPC
UniProt ID: (Human) P23582, (Mouse) Q61839, (Rat) P55207
Entrez Gene ID: (Human) 4880, (Mouse) 18159, (Rat) 114593
Molecular Function: peptide hormone signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.