Invitrogen
Manufacturer Code:PA528287
Catalog # PIPA528287
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit fragment corresponding to a region within amino acids 67 and 292 of Human CNOT8 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: CAF1 CALIF POP2 hCAF1 CAF1-like protein CAF2 CALIFp CCR4-NOT transcription complex subunit 8 CCR4-associated factor 8 PGK promoter directed over production; CAF1-like protein; Caf1b; CAF2; CALIFp; CCR4-associated factor 8; CCR4-NOT transcription complex subunit 8; CCR4-NOT transcription complex, subunit 8; PGK promoter directed over production
Gene Aliases: CAF1; Caf1b; CALIF; CNOT8; hCAF1; POP2
UniProt ID: (Human) Q9UFF9
Entrez Gene ID: (Human) 9337
Molecular Function: transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.