Proteintech
Manufacturer Code:107521AP150UL
Catalog # 50-172-6567
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | CNOT8 Fusion Protein Ag1201 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: CAF1 CAF1 like protein CAF2 CALIF CALIFp CCR4 associated factor 8 CNOT8 hCAF1 POP2; CAF1-like protein; Caf1b; CAF2; CALIFp; CCR4-associated factor 8; CCR4-NOT transcription complex subunit 8; CCR4-NOT transcription complex, subunit 8; PGK promoter directed over production
Gene Aliases: 1500015I04Rik; 1810022F04Rik; AA536816; AU015770; AU043059; CAF1; Caf1b; CALIF; CNOT8; hCAF1; POP2
UniProt ID: (Human) Q9UFF9, (Mouse) Q9D8X5
Entrez Gene ID: (Human) 9337, (Mouse) 69125, (Rat) 363603
Molecular Function:
transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.