Invitrogen
Manufacturer Code:PA567055
Catalog # PIPA567055
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit Human CNOT7 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: BTG1-binding factor 1; CAF-1; CAF1 hCAF-1 BTG1 binding factor 1 BTG1-binding factor 1 CAF-1 CCR4-NOT transcription complex subunit 7 CCR4-associated factor 1 carbon catabolite repressor protein (CCR4)-associative factor 1; Caf1a; carbon catabolite repressor protein (CCR4)-associative factor 1; CCR4-associated factor 1; CCR4-NOT transcription complex subunit 7
Gene Aliases: CAF-1; CAF1; Caf1a; CNOT7; hCAF-1
UniProt ID: (Human) Q9UIV1
Entrez Gene ID: (Human) 29883
Molecular Function:
transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.