Proteintech
Manufacturer Code:141021AP150UL
Catalog # 50-172-9211
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | CNOT7 Fusion Protein Ag5235 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: BTG1 binding factor 1 CAF 1 CAF1 CCR4 associated factor 1 CNOT7 hCAF 1; BTG1-binding factor 1; CAF-1; Caf1a; carbon catabolite repressor protein (CCR4)-associative factor 1; catabolite repressor protein (CCR4)-associative factor 1; CCR4-associated factor 1; CCR4-NOT transcription complex subunit 7; POP2 homolog
Gene Aliases: AU022737; CAF-1; CAF1; Caf1a; CNOT7; hCAF-1; Pop2
UniProt ID: (Human) Q9UIV1, (Mouse) Q60809
Entrez Gene ID: (Human) 29883, (Mouse) 18983, (Rat) 306492
Molecular Function:
transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.