Invitrogen
Manufacturer Code:PA5100444
Catalog # PIPA5100444
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthesized peptide derived from human CNOT6 corresponding to a region within C-terminal amino acids. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: carbon catabolite repression 4 protein; carbon catabolite repression 4 protein carbon catabolite repressor protein 4 homolog Ccr4 CCR4 carbon catabolite repression 4-like Ccr4a CCR4-NOT transcription complex subunit 6 CCR4-NOT transcription complex subunit 6 Cnot6 cytoplasmic deadenylase Kiaa1194 RGD1310783; Carbon catabolite repressor protein 4 homolog; CCR4 carbon catabolite repression 4-like; CCR4-NOT transcription complex subunit 6; CCR4-NOT transcription complex, subunit 6; Cytoplasmic deadenylase
Gene Aliases: CCR4; Ccr4a; Cnot6; Kiaa1194; RGD1310783
UniProt ID: (Rat) Q6AXU9, (Mouse) Q8K3P5
Entrez Gene ID: (Human) 57472, (Rat) 287249, (Mouse) 104625
Molecular Function: RNA binding protein exoribonuclease nuclease nucleic acid binding
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.