Novus Biologicals
Manufacturer Code:NBP231892
Catalog # NBP231892
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | This antibody was developed against a recombinant protein corresponding to amino acids: ALLQINTSWHTLRHELISTLMPIFLGNHPNSAIILHYAWHGQGQSPSIRQLIMHAMAEWYMRGEQYDQAKLSRILDVAQDLKALSMLLNGT |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: AD-005 adrenal gland protein AD-005 CCR4-associated factor 1 CCR4-NOT transcription complex subunit 1 CCR4-NOT transcription complex subunit 1 CDC39DKFZp686E0722 hNOT1 KIAA1007DKFZp686O168 Negative regulator of transcription subunit 1 homolog NOT1 (negative regulator of transcription 1 yeast) homolog NOT1FLJ90644 NOT1HFLJ36492; adrenal gland protein AD-005; CCR4-associated factor 1; CCR4-NOT transcription complex subunit 1; CCR4-NOT transcription complex, subunit 1; Negative regulator of transcription subunit 1 homolog; NOT1 (negative regulator of transcription 1, yeast) homolog; NOT1H
Gene Aliases: AD-005; CDC39; CNOT1; KIAA1007; NOT1; NOT1H
UniProt ID: (Human) A5YKK6
Entrez Gene ID: (Human) 23019
Molecular Function:
transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.