Proteintech
Manufacturer Code:142761AP150UL
Catalog # 50-557-559
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | CNOT1 Fusion Protein Ag5623 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: AD 005 CCR4 associated factor 1 CDC39 CNOT1 hNOT1 KIAA1007 NOT1 NOT1H; adrenal gland protein AD-005; CCR4-associated factor 1; CCR4-NOT transcription complex subunit 1; CCR4-NOT transcription complex, subunit 1; Negative regulator of transcription subunit 1 homolog; NOT1 (negative regulator of transcription 1, yeast) homolog; NOT1H
Gene Aliases: 6030411K04Rik; AA815922; AD-005; CDC39; CNOT1; D830048B13; KIAA1007; NOT1; NOT1H; RGD1308009
UniProt ID: (Human) A5YKK6, (Mouse) Q6ZQ08
Entrez Gene ID: (Human) 23019, (Rat) 291841, (Mouse) 234594
Molecular Function:
transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.