Proteintech
Manufacturer Code:140661AP150UL
Catalog # 50-172-9185
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | CNNM4 Fusion Protein Ag5121 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: ACDP4 CNNM4 cyclin M4 KIAA1592 Metal transporter CNNM4; ancient conserved domain protein 4; Ancient conserved domain-containing protein 4; cyclin M4; Cyclin-M4; mACDP4; Metal transporter CNNM4
Gene Aliases: 5430430O18Rik; ACDP4; CNNM4; KIAA1592
UniProt ID: (Human) Q6P4Q7, (Mouse) Q69ZF7, (Rat) P0C588
Entrez Gene ID: (Human) 26504, (Mouse) 94220, (Rat) 363216
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.