Proteintech
Manufacturer Code:115091AP150UL
Catalog # 50-172-7223
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | CNN3 Fusion Protein Ag2068 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: acidic calponin h3; Calponin 3 calponin 3 acidic Calponin acidic isoform CNN3; calponin 3, acidic; Calponin, acidic isoform; Calponin, non-muscle isoform; Calponin-3; dJ639P13.2.2 (acidic calponin 3)
Gene Aliases: 1600014M03Rik; C85854; Calpo3; CNN3
UniProt ID: (Human) Q15417, (Mouse) Q9DAW9, (Rat) P37397
Entrez Gene ID: (Human) 1266, (Mouse) 71994, (Rat) 54321
Molecular Function: actin family cytoskeletal protein cytoskeletal protein non-motor actin binding protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.