Proteintech
Manufacturer Code:662441IG150UL
Catalog # 50-173-6677
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 1G10F5 |
Immunogen | CNN2 Fusion Protein Ag23594 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: calponin 2 Calponin H2 smooth muscle CNN2 Neutral calponin; Calponin H2, smooth muscle; Calponin-2; h2-calponin; Neutral calponin
Gene Aliases: Calponin-2; CNN2
UniProt ID: (Human) Q99439
Entrez Gene ID: (Human) 1265, (Pig) 397565
Molecular Function:
actin family cytoskeletal protein
cytoskeletal protein
non-motor actin binding protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.