Proteintech
Manufacturer Code:210731AP150UL
Catalog # 50-173-3242
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | CNN2 Fusion Protein Ag15225 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: calponin 2; calponin 2 Calponin H2 smooth muscle CNN2 Neutral calponin; Calponin H2, smooth muscle; Calponin-2; h2-calponin; Neutral calponin
Gene Aliases: AA408047; AI324678; Calpo2; CNN2
UniProt ID: (Human) Q99439, (Mouse) Q08093
Entrez Gene ID: (Human) 1265, (Mouse) 12798, (Rat) 690976
Molecular Function: actin family cytoskeletal protein cytoskeletal protein non-motor actin binding protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.