Invitrogen
Manufacturer Code:PA5113040
Catalog # PIPA5113040
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant Human Protein cornichon homolog 2 protein (94-138AA) |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: CNIH-2; CNIH2 CNIH-2 CNIL Cornichon family AMPA receptor auxiliary protein 2 cornichon homolog 2 Cornichon-like protein Protein cornichon homolog 2; Cornichon family AMPA receptor auxiliary protein 2; cornichon homolog 2; Cornichon-like protein; Protein cornichon homolog 2
Gene Aliases: CNIH-2; CNIH2; CNIL
UniProt ID: (Human) Q6PI25, (Mouse) O35089
Entrez Gene ID: (Human) 254263, (Mouse) 12794
Molecular Function: membrane traffic protein membrane-bound signaling molecule signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.