Invitrogen
Manufacturer Code:PA5101543
Catalog # PIPA5101543
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthesized peptide derived from human CNGB1 corresponding to a region within the internal amino acids. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: CNCG2 CNCG3L CNCG4 cng channel CNG channel 4 CNG channel beta-1 CNG4 CNG-4 CNGB1 CNGB1B cyclic nucleotide gated channel beta 1 cyclic nucleotide-gated cation channel 4 Cyclic nucleotide-gated cation channel beta-1 Cyclic nucleotide-gated cation channel gamma cyclic nucleotide-gated cation channel modulatory subunit cyclic nucleotide-gated channel beta subunit 1 cyclic nucleotide-gated channel beta-1 GAR1 GARP GARP2 Glutamic acid-rich protein glutamic-acid-rich protein Gm1959 RCNC2 RCNCb RCNCbeta RP45; CNG channel 4; CNG channel beta-1; CNG-4; Cyclic nucleotide-gated cation channel 4; Cyclic nucleotide-gated cation channel beta-1; Cyclic nucleotide-gated cation channel gamma; Cyclic nucleotide-gated cation channel modulatory subunit; cyclic nucleotide-gated channel beta subunit 1; Cyclic nucleotide-gated channel beta-1; GARP; Glutamic acid-rich protein; glutamic-acid-rich protein
Gene Aliases: CNCG2; CNCG3L; CNCG4; CNG4; CNGB1; CNGB1B; GAR1; GARP; GARP2; Gm1959; RCNC2; RCNCb; RCNCbeta; RP45
UniProt ID: (Human) Q14028
Entrez Gene ID: (Human) 1258, (Rat) 83686, (Mouse) 333329
Molecular Function: cyclic nucleotide-gated ion channel ion channel ligand-gated ion channel potassium channel receptor transporter voltage-gated ion channel voltage-gated potassium channel
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.