Invitrogen
Manufacturer Code:PA577616
Catalog # PIPA577616
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Peptide (C) SQDGKVKTTESTPPAPTK corresponding to amino acid residues 2-19 of rat CNGA4 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: CNCalpha4, CNG5; CNG channel alpha-4; CNG-4; CNG4; cyclic nucleotide gated cation channel; cyclic nucleotide-gated cation channel alpha 4; Cyclic nucleotide-gated cation channel alpha-4; Cyclic nucleotide-gated channel alpha-4; Cyclic nucleotide-gated olfactory channel subunit OCNC2; OCNC2; olfactory cyclic nucleotide-gated channel
Gene Aliases: Cgn2; Cnga4; rOCNC2
UniProt ID: (Mouse) Q3UW12, (Rat) Q64359
Entrez Gene ID: (Mouse) 233649, (Rat) 85258
Molecular Function: cyclic nucleotide-gated ion channel ion channel ligand-gated ion channel potassium channel receptor transporter voltage-gated ion channel voltage-gated potassium channel
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.