Invitrogen
Manufacturer Code:PA5114327
Catalog # PIPA5114327
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide corresponding to N terminal Rat CNGA4. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Cgn2 CNCA2 CNCalpha4 CNG5 cng channel CNG channel alpha-4 CNG4 CNG-4 CNG5 CNGA4 CNGB2 cyclic nucleotide gated cation channel cyclic nucleotide gated channel alpha 4 cyclic nucleotide gated channel beta 2 cyclic nucleotide-gated cation channel alpha 4 Cyclic nucleotide-gated cation channel alpha-4 cyclic nucleotide-gated channel alpha-4 Cyclic nucleotide-gated olfactory channel subunit OCNC2 OCNC2 OCNCb OCNCBETA olfactory cyclic nucleotide-gated channel rOCNC2; CNCalpha4, CNG5; CNG channel alpha-4; CNG-4; CNG4; cyclic nucleotide gated cation channel; cyclic nucleotide gated channel beta 2; cyclic nucleotide-gated cation channel alpha 4; Cyclic nucleotide-gated cation channel alpha-4; Cyclic nucleotide-gated channel alpha-4; Cyclic nucleotide-gated olfactory channel subunit OCNC2; olfactory cyclic nucleotide-gated channel
Gene Aliases: Cgn2; CNCA2; CNG-4; CNG4; CNG5; CNGA4; CNGB2; OCNC2; OCNCb; OCNCBETA; rOCNC2
UniProt ID: (Human) Q8IV77, (Mouse) Q3UW12, (Rat) Q64359
Entrez Gene ID: (Human) 1262, (Mouse) 233649, (Rat) 85258
Molecular Function: cyclic nucleotide-gated ion channel ion channel ligand-gated ion channel potassium channel receptor transporter voltage-gated ion channel voltage-gated potassium channel
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.