Proteintech
Manufacturer Code:216571AP150UL
Catalog # 50-173-3542
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | CNGA3 Fusion Protein Ag16385 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: ACHM2 CCNC1 CCNCa CCNCalpha CNCG3 CNG 3 CNG channel alpha 3 CNG3 CNGA3; CNG channel alpha-3; CNG-3; cone photoreceptor cGMP-gated channel alpha subunit; Cone photoreceptor cGMP-gated channel subunit alpha; Cyclic nucleotide-gated cation channel alpha-3; Cyclic nucleotide-gated channel alpha-3
Gene Aliases: ACHM2; CCNC1; CCNCa; CCNCalpha; CNCG3; CNG3; CNGA3
UniProt ID: (Human) Q16281, (Mouse) Q9JJZ8
Entrez Gene ID: (Human) 1261, (Mouse) 12790
Molecular Function: cyclic nucleotide-gated ion channel ion channel ligand-gated ion channel potassium channel receptor transporter voltage-gated ion channel voltage-gated potassium channel
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.