Invitrogen
Manufacturer Code:PA577604
Catalog # PIPA577604
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Frozen) (IHC (F)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Peptide (C)KQNHEDDYLSDGINTPEP corresponding to amino acid residues 643-660 of rat CNGA2. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: CNG channel alpha-2; CNG-2; CNG2; Cyclic Nucleotide Gated Cation Channel 2CNG2 OCNC1 Olfactory Cyclic Nucleotide Gated Channel 1; cyclic nucleotide gated channel 4; Cyclic nucleotide-gated cation channel 2; Cyclic nucleotide-gated channel alpha-2; Cyclic nucleotide-gated olfactory channel; Cyclic nucleotide-gated olfactory channel subunit OCNC1; cyclic-nucleotide-gated cation channel 2
Gene Aliases: CNCA; CNCA1; CNCG2; Cncg4; CNG2; CNGA2; OCNC1; OCNCa; OCNCALPHA; OLFCH
UniProt ID: (Human) Q16280, (Rat) Q00195, (Mouse) Q62398
Entrez Gene ID: (Human) 1260, (Rat) 25411, (Mouse) 12789
Molecular Function: cyclic nucleotide-gated ion channel ion channel ligand-gated ion channel potassium channel receptor transporter voltage-gated ion channel voltage-gated potassium channel
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.