Invitrogen
Manufacturer Code:PA5111881
Catalog # PIPA5111881
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Rat |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Peptide corresponding to amino acid residues 41-54 of rat CNGA1 (Intracellular N-terminus) |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: cGMP-gated cation channel alpha-1; cGMP-gated cation channel alpha-1 Cncg Cncg1 CNG channel alpha-1 CNG1 CNG-1 CNGA1 Cyclic nucleotide-gated cation channel 1 Cyclic nucleotide-gated channel alpha-1 Cyclic nucleotide-gated channel photoreceptor HCN RCNC1 RCNCa RCNCalpha Rod photoreceptor cGMP-gated channel subunit alpha RP49; CNG channel alpha-1; CNG-1; CNG1; Cyclic nucleotide-gated cation channel 1; Cyclic nucleotide-gated channel alpha-1; Cyclic nucleotide-gated channel, photoreceptor; Rod photoreceptor cGMP-gated channel subunit alpha
Gene Aliases: Cncg; Cncg1; Cnga1; HCN
UniProt ID: (Rat) Q62927
Entrez Gene ID: (Rat) 85259
Molecular Function: cyclic nucleotide-gated ion channel ion channel ligand-gated ion channel potassium channel receptor transporter voltage-gated ion channel voltage-gated potassium channel
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.