Invitrogen
Manufacturer Code:PA5101516
Catalog # PIPA5101516
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthesized peptide derived from human CNGA1 corresponding to a region within the internal amino acids. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: cGMP-gated cation channel alpha-1; cGMP-gated cation channel alpha-1 CNCG CNCG1 cng channel CNG channel alpha-1 CNG1 CNG-1 Cnga1 cyclic nucleotide gated channel alpha 1 cyclic nucleotide gated channel cGMP gated Cyclic nucleotide-gated cation channel 1 Cyclic nucleotide-gated channel alpha-1 cyclic nucleotide-gated channel photoreceptor HCN interleukin-1 homologue RCNC1 RCNCa RCNCalpha Rod photoreceptor cGMP-gated channel subunit alpha RP49; CNG channel alpha-1; CNG-1; CNG1; cyclic nucleotide gated channel, cGMP gated; Cyclic nucleotide-gated cation channel 1; Cyclic nucleotide-gated channel alpha-1; Cyclic nucleotide-gated channel, photoreceptor; interleukin-1 homologue; Rod photoreceptor cGMP-gated channel subunit alpha
Gene Aliases: CNCG; CNCG1; CNG-1; CNG1; CNGA1; HCN; RCNC1; RCNCa; RCNCalpha; RP49
UniProt ID: (Human) P29973, (Mouse) P29974, (Rat) Q62927
Entrez Gene ID: (Human) 1259, (Mouse) 12788, (Rat) 85259
Molecular Function: cyclic nucleotide-gated ion channel ion channel ligand-gated ion channel potassium channel receptor transporter voltage-gated ion channel voltage-gated potassium channel
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.